5-Bromo-2,4-dichloropyrimidine 99%
Supplier: Apollo Scientific
Danger
OR9965-100G OR9965-25G OR9965-5G
APOSOR9965-100GEA 65.4 EUR
APOSOR9965-100G APOSOR9965-25G APOSOR9965-5G
5-Bromo-2,4-dichloropyrimidine 99%
5-Bromo-2,4-dichloropyrimidine
Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 128 °C/5mmHg
Melting Pt: 29…30 °C
Density: 1.781 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2922
ADR: 8+6.1 III