2-Bromo-4-phenylpyridine 97+%

Supplier: Apollo Scientific
Warning

OR9966-10G OR9966-1G OR9966-5G
APOSOR9966-10GEA 739 EUR
APOSOR9966-10G APOSOR9966-1G APOSOR9966-5G
2-Bromo-4-phenylpyridine 97+%
2-Bromo-4-phenylpyridine
Formula: C₁₁H₈BrN
MW: 234.0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235159
CAS Number: 54151-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR