4,6-Dichloro-3-(3-chlorophenyl)pyridazine 95+%
Supplier: Apollo Scientific
OR9970-1G OR9970-5G
APOSOR9970-1GEA 260 EUR
APOSOR9970-1G APOSOR9970-5G
4,6-Dichloro-3-(3-chlorophenyl)pyridazine 95+%
4,6-Dichloro-3-(3-chlorophenyl)pyridazine
Formula: C₁₀H₅Cl₃N₂
Storage Temperature: Ambient
MDL Number: MFCD06800549
CAS Number: 849021-04-1