4-Chloro-6,7-dimethoxyquinazoline 98%
Supplier: Apollo Scientific
Warning
OR9985-1G OR9985-5G OR9985-25G
APOSOR9985-1GEA 62.5 EUR
APOSOR9985-1G APOSOR9985-5G APOSOR9985-25G
4-Chloro-6,7-dimethoxyquinazoline 98%
4-Chloro-6,7-dimethoxyquinazoline
Formula: C₁₀H₉ClN₂O₂
MW: 224.65 g/mol
Melting Pt: 284…287 °C
Storage Temperature: Ambient
MDL Number: MFCD01570172
CAS Number: 13790-39-1