5-Chloro-3-methyl-1-phenyl-1H-pyrazole 98%
Supplier: Apollo Scientific
Synonyms: 5-Chloro-3-methyl-1-phenyl-1H-pyrazole
OR9991-1G OR9991-5G
APOSOR9991-1GEA 50.9 EUR
APOSOR9991-1G APOSOR9991-5G
5-Chloro-3-methyl-1-phenyl-1H-pyrazole 98%
5-Chloro-3-methyl-1-phenyl-1H-pyrazole
Formula: C₁₀H₉ClN₂
MW: 192.65 g/mol
Boiling Pt: 114…115 °C/4mmHg
Storage Temperature: Ambient
MDL Number: MFCD00067853
CAS Number: 1131-17-5