3-Chloro-2,6-difluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Chloro-2,6-difluorobenzene-1-carbonyl chloride
PC0001-1G PC0001-5G
APOSPC0001-1GEA 29 EUR
APOSPC0001-1G APOSPC0001-5G
3-Chloro-2,6-difluorobenzoyl chloride 98%
3-Chloro-2,6-difluorobenzoyl chloride
Formula: C₇H₂Cl₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD01631324
CAS Number: 261762-43-0
UN: 3265
ADR: 8,II