ω-Chloro-2,4-difluoroacetophenone 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone
PC0002-100G PC0002-25G PC0002-5G
APOSPC0002-100GEA 78.3 EUR
APOSPC0002-100G APOSPC0002-25G APOSPC0002-5G
ω-Chloro-2,4-difluoroacetophenone 98%
ω-Chloro-2,4-difluoroacetophenone
Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Melting Pt: 44…48 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013252
CAS Number: 51336-94-8
UN: 3261
ADR: 8,II