3-Chloro-2-fluorobenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC0044-1G PC0044-25G PC0044-5G
APOSPC0044-1GEA 21.9 EUR
APOSPC0044-1G APOSPC0044-25G APOSPC0044-5G
3-Chloro-2-fluorobenzoyl chloride 97%
3-Chloro-2-fluorobenzoyl chloride
Formula: C₇H₃Cl₂FO
MW: 193 g/mol
Boiling Pt: 202…203 °C
Density: 1.451 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD01631548
CAS Number: 85345-76-2
UN: 2920
ADR: 8+3,II