2-Chloro-4-fluorobenzyl alcohol 98%
Supplier: Apollo Scientific
Warning
PC0048-25G PC0048-5G PC0048-100G
APOSPC0048-25GEA 68.3 EUR
APOSPC0048-25G APOSPC0048-5G APOSPC0048-100G
2-Chloro-4-fluorobenzyl alcohol 98%
2-Chloro-4-fluorobenzyl alcohol
Formula: C₇H₆ClFO
Melting Pt: 44…46 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00236024
CAS Number: 208186-84-9