1-Chloro-3-fluoro-4-iodobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-1-fluoro-2-iodobenzene, 4-Chloro-2-fluoro-1-iodobenzene, 4-Chloro-2-fluoroiodobenzene
PC0053-100G PC0053-25G PC0053-500G
APOSPC0053-100GEA 65.4 EUR
APOSPC0053-100G APOSPC0053-25G APOSPC0053-500G
1-Chloro-3-fluoro-4-iodobenzene 97%
1-Chloro-3-fluoro-4-iodobenzene
Formula: C₆H₃ClFI
MW: 256.45 g/mol
Boiling Pt: 107 °C/18mmHg
Melting Pt: 17…19 °C
Storage Temperature: Ambient
MDL Number: MFCD00079729
CAS Number: 6797-79-1