5-Chloro-2-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC0082-100G PC0082-25G PC0082-5G
APOSPC0082-100GEA 69.6 EUR
APOSPC0082-100G APOSPC0082-25G APOSPC0082-5G
5-Chloro-2-fluoroaniline 98%
5-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 102…105 °C/20mmHg
Melting Pt: 20…22 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00069416
CAS Number: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III