6-Chloro-2-fluoro-3-methylbenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC0094-25G PC0094-5G PC0094-1G
APOSPC0094-25GEA 319 EUR
APOSPC0094-25G APOSPC0094-5G APOSPC0094-1G
6-Chloro-2-fluoro-3-methylbenzaldehyde 98%
6-Chloro-2-fluoro-3-methylbenzaldehyde
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 249 °C
Density: 1.3 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631370
CAS Number: 286474-59-7