6-Chloro-2-fluoro-3-methylbenzyl bromide 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Bromomethyl)-1-chloro-3-fluoro-4-methylbenzene
PC0116-10G PC0116-1G PC0116-5G
APOSPC0116-10GEA 284 EUR
APOSPC0116-10G APOSPC0116-1G APOSPC0116-5G
6-Chloro-2-fluoro-3-methylbenzyl bromide 98%
6-Chloro-2-fluoro-3-methylbenzyl bromide
Formula: C₈H₇BrClF
MW: 237.51 g/mol
Boiling Pt: 130 °C/3mmHg
Melting Pt: 52…58 °C
Storage Temperature: Ambient
MDL Number: MFCD01631373
CAS Number: 261762-88-3
UN: 1759
ADR: 8,III