1-Chloro-3,4-dibromo-5-fluorobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dibromo-5-chloro-3-fluorobenzene, 5-Chloro-2,3-dibromo-1-fluorobenzene, 5-Chloro-1,2-dibromo-3-fluorobenzene
PC0121-25G PC0121-100G
APOSPC0121-25GEA 75.4 EUR
APOSPC0121-25G APOSPC0121-100G
1-Chloro-3,4-dibromo-5-fluorobenzene 98%
1-Chloro-3,4-dibromo-5-fluorobenzene
Formula: C₆H₂Br₂ClF
MW: 288.34 g/mol
Boiling Pt: 242
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00143441
CAS Number: 208186-78-1