3-Chloro-2,4-difluorobenzoic acid 97%
Supplier: Apollo Scientific
Warning
PC0145-25G PC0145-5G PC0145-100G
APOSPC0145-25GEA 65.4 EUR
APOSPC0145-25G APOSPC0145-5G APOSPC0145-100G
3-Chloro-2,4-difluorobenzoic acid 97%
3-Chloro-2,4-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Melting Pt: 169…177 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631387
CAS Number: 154257-75-7