3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC0146-25G PC0146-5G PC0146-1G
APOSPC0146-25GEA 484 EUR
APOSPC0146-25G APOSPC0146-5G APOSPC0146-1G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde 98%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₃ClF₄O
MW: 226.56 g/mol
Boiling Pt: 214 °C
Density: 1.53 g/cm³ (25 °C)
Flash Pt: 104.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01631343
CAS Number: 186517-29-3