3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride 95%
Supplier: Apollo Scientific
Danger
PC0152-1G PC0152-25G PC0152-5G
APOSPC0152-1GEA 26.1 EUR
APOSPC0152-1G APOSPC0152-25G APOSPC0152-5G
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride 95%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoyl chloride
Formula: C₈H₂Cl₂F₄O
MW: 261 g/mol
Boiling Pt: 230 °C
Melting Pt: 85
Storage Temperature: Ambient
MDL Number: MFCD01631511
CAS Number: 261763-03-5
UN: 3265
ADR: 8 II