4-Trifluoromethyl-L-phenylalanine 97%
Supplier: Apollo Scientific
Warning
Synonyms: H-Phe(4-CF3)-OH, 4-(Trifluoromethyl)-L-phenylalanine
PC01528-1G PC01528-5G
APOSPC01528-1GEA 102 EUR
APOSPC01528-1G APOSPC01528-5G
4-Trifluoromethyl-L-phenylalanine 97%
4-Trifluoromethyl-L-phenylalanine
Formula: C₁₀H₁₀F₃NO₂
MW: 233.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044946
CAS Number: 114926-38-4