(3-Bromo-5-(trifluoromethyl)phenyl)methanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-5-(trifluoromethyl)benzyl alcohol
PC01529-1G PC01529-25G PC01529-5G
APOSPC01529-1GEA 21.9 EUR
APOSPC01529-1G APOSPC01529-25G APOSPC01529-5G
(3-Bromo-5-(trifluoromethyl)phenyl)methanol 95%
(3-Bromo-5-(trifluoromethyl)phenyl)methanol
Formula: C₈H₆BrF₃O
MW: 255.0300 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD09835207
CAS Number: 172023-97-1