2-Chloro-5-(trifluoromethyl)benzoyl chloride
Supplier: Apollo Scientific
Danger
PC0188-1G PC0188-5G
APOSPC0188-1GEA 23.2 EUR
APOSPC0188-1G APOSPC0188-5G
2-Chloro-5-(trifluoromethyl)benzoyl chloride
2-Chloro-5-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃Cl₂F₃O
Boiling Pt: 59…61 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD01631634
CAS Number: 657-05-6
UN: 3265
ADR: 8,II