2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-4-carboxylic acid
PC0221-1G PC0221-5G
APOSPC0221-1GEA 21.9 EUR
APOSPC0221-1G APOSPC0221-5G
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid 98%
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid
Formula: C₈H₄F₂O₄
MW: 202.12 g/mol
Melting Pt: 200…204 °C
Storage Temperature: Ambient
MDL Number: MFCD01631473
CAS Number: 126120-85-2
UN: 3077
ADR: 9+EHS,III