2,3-Difluoro-4-methoxyphenol
Supplier: Apollo Scientific
Danger
Synonyms: 2,3-Difluor-4-hydroxyanisole
PC0226-1G PC0226-5G
APOSPC0226-1GEA 21.9 EUR
APOSPC0226-1G APOSPC0226-5G
2,3-Difluoro-4-methoxyphenol
2,3-Difluoro-4-methoxyphenol
Formula: C₇H₆F₂O₂
MW: 160.12 g/mol
Melting Pt: 103…106 °C
Storage Temperature: Ambient
MDL Number: MFCD01631505
CAS Number: 261763-29-5