2,6-Difluoro-3-methylbenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Difluoro-3-tolualdehyde, 2,6-Difluoro-m-tolualdehyde, 2,6-Difluoro-m-tolylaldehyde, 2,6-Difluoro-3-tolylaldehyde
PC0239-25G PC0239-5G PC0239-100G
APOSPC0239-25GEA 389 EUR
APOSPC0239-25G APOSPC0239-5G APOSPC0239-100G
2,6-Difluoro-3-methylbenzaldehyde
2,6-Difluoro-3-methylbenzaldehyde
Formula: C₈H₆F₂O
MW: 156.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631333
CAS Number: 261763-34-2