2,3-Difluoro-p-toluic acid 97%
Supplier: Apollo Scientific
Warning
PC0245-5G PC0245-1G
APOSPC0245-5GEA 94.4 EUR
APOSPC0245-5G APOSPC0245-1G
2,3-Difluoro-p-toluic acid 97%
2,3-Difluoro-p-toluic acid
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Melting Pt: 199…201 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631643
CAS Number: 261763-37-5