2,6-Difluoro-3-methylbenzylamine 98%
Supplier: Apollo Scientific
PC0252-10G PC0252-5G PC0252-1G
APOSPC0252-10GEA 360 EUR
APOSPC0252-10G APOSPC0252-5G APOSPC0252-1G
2,6-Difluoro-3-methylbenzylamine 98%
2,6-Difluoro-3-methylbenzylamine
Formula: C₈H₉F₂N
MW: 157.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631410
CAS Number: 261763-42-2