5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulphonyl chloride 95%
Supplier: Apollo Scientific
Danger
PC0273-1G PC0273-250MG
APOSPC0273-1GEA 241 EUR
APOSPC0273-1G APOSPC0273-250MG
5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulphonyl chloride 95%
5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulphonyl chloride
Formula: C₉H₆ClF₃N₂O₂S₂
Melting Pt: 65…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00830811
CAS Number: 230295-11-1
UN: 1759
ADR: 8,III