2,3,6-Trifluorocinnamic acid 98%
Supplier: Apollo Scientific
Synonyms: 3-(2,3,6-Trifluorophenyl)acrylic acid
PC0275-5G PC0275-1G
APOSPC0275-5GEA 144 EUR
APOSPC0275-5G APOSPC0275-1G
2,3,6-Trifluorocinnamic acid 98%
2,3,6-Trifluorocinnamic acid
Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061213
CAS Number: 230295-12-2