2-(3-Trifluoromethylphenyl)malondialdehyde 95+%
Supplier: Apollo Scientific
Warning
PC0314-1G PC0314-5G PC0314-25G
APOSPC0314-1GEA 185 EUR
APOSPC0314-1G APOSPC0314-5G APOSPC0314-25G
2-(3-Trifluoromethylphenyl)malondialdehyde 95+%
2-(3-Trifluoromethylphenyl)malondialdehyde
Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD03424496
CAS Number: 493036-49-0