5-Bromo-2-fluoro-m-xylene 99%
Supplier: Apollo Scientific
Warning
PC0327-1G PC0327-25G PC0327-5G
APOSPC0327-1GEA 21.9 EUR
APOSPC0327-1G APOSPC0327-25G APOSPC0327-5G
5-Bromo-2-fluoro-m-xylene 99%
5-Bromo-2-fluoro-m-xylene
Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 95 °C/22mmHg
Storage Temperature: Ambient
MDL Number: MFCD01320701
CAS Number: 99725-44-7