3'-Fluoro-4'-methylacetophenone 98%
Supplier: Apollo Scientific
Warning
PC0338-25G PC0338-5G PC0338-1G
APOSPC0338-25GEA 393 EUR
APOSPC0338-25G APOSPC0338-5G APOSPC0338-1G
3'-Fluoro-4'-methylacetophenone 98%
3'-Fluoro-4'-methylacetophenone
Formula: C₉H₉FO
Melting Pt: 31…34 °C
Storage Temperature: Ambient
MDL Number: MFCD01631411
CAS Number: 42444-14-4