4,5-Difluoro-o-phenylenediamine
Supplier: Apollo Scientific
Warning
Synonyms: 4,5-Difluoro-1,2-phenylenediamine, 4,5-Difluorobenzene-1,2-diamine, 1,2-Diamino-4,5-difluorobenzene
PC0342-25G PC0342-100G PC0342-5G
APOSPC0342-25GEA 109 EUR
APOSPC0342-25G APOSPC0342-100G APOSPC0342-5G
4,5-Difluoro-o-phenylenediamine
4,5-Difluoro-o-phenylenediamine
Formula: C₆H₆F₂N₂
MW: 144.12 g/mol
Melting Pt: 131…132 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061131
CAS Number: 76179-40-3