4-Fluoro-m-toluic acid
Supplier: Apollo Scientific
Warning
PC0345-25G PC0345-5G
APOSPC0345-25GEA 24.8 EUR
APOSPC0345-25G APOSPC0345-5G
4-Fluoro-m-toluic acid
4-Fluoro-m-toluic acid
Formula: C₈H₇FO₂
Melting Pt: 164…169 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631424
CAS Number: 403-15-6