2,4,5-Trifluorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4,5-Trifluorobenzene-1-sulphonyl chloride
PC0349-100G PC0349-25G PC0349-5G
APOSPC0349-100GEA 161 EUR
APOSPC0349-100G APOSPC0349-25G APOSPC0349-5G
2,4,5-Trifluorobenzenesulphonyl chloride 98%
2,4,5-Trifluorobenzenesulphonyl chloride
Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
Density: 1.657 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00077600
CAS Number: 220227-21-4
UN: 3265
ADR: 8,II