4-Chloro-3-fluoroaniline 98%
Supplier: Apollo Scientific
Danger
PC0361-25G PC0361-5G
APOSPC0361-25GEA 55.1 EUR
APOSPC0361-25G APOSPC0361-5G
4-Chloro-3-fluoroaniline 98%
4-Chloro-3-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 226 °C
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD01090987
CAS Number: 367-22-6
EINECS: 206-683-3
UN: 2811
ADR: 6.1,III