2-Chloro-5-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC0364-100G PC0364-25G PC0364-500G
APOSPC0364-100GEA 71.2 EUR
APOSPC0364-100G APOSPC0364-25G APOSPC0364-500G
2-Chloro-5-fluorobenzoic acid 98%
2-Chloro-5-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00156967
CAS Number: 2252-50-8