5-Fluoro-2-methylbenzyl bromide 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Bromomethyl)-4-fluoro-1-methylbenzene
PC0365-25G PC0365-5G PC0365-1G
APOSPC0365-25GEA 179 EUR
APOSPC0365-25G APOSPC0365-5G APOSPC0365-1G
5-Fluoro-2-methylbenzyl bromide 97%
5-Fluoro-2-methylbenzyl bromide
Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 84…85 °C/8mmHg
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631528
CAS Number: 261951-71-7
UN: 3265
ADR: 8 II