2-Fluoro-5-(trifluoromethyl)anisole 98%
Supplier: Apollo Scientific
PC0381-5G PC0381-1G
APOSPC0381-5GEA 106 EUR
APOSPC0381-5G APOSPC0381-1G
2-Fluoro-5-(trifluoromethyl)anisole 98%
2-Fluoro-5-(trifluoromethyl)anisole
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Boiling Pt: 151
Melting Pt: -15
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD01631564
CAS Number: 261951-78-4