3-Fluoro-2-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Warning
PC0390-1G PC0390-5G PC0390-10G
APOSPC0390-1GEA 21.9 EUR
APOSPC0390-1G APOSPC0390-5G APOSPC0390-10G
3-Fluoro-2-(trifluoromethyl)benzonitrile 98%
3-Fluoro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631566
CAS Number: 261951-81-9
ADR: 6.1,III