2-Bromo-α,α,α,6-tetrafluorotoluene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-3-fluoro-2-(trifluoromethyl)benzene, 2-Bromo-6-fluorobenzotrifluoride
PC0395-100G PC0395-25G PC0395-500G
APOSPC0395-100GEA 284 EUR
APOSPC0395-100G APOSPC0395-25G APOSPC0395-500G
2-Bromo-α,α,α,6-tetrafluorotoluene 97%
2-Bromo-α,α,α,6-tetrafluorotoluene
Formula: C₇H₃BrF₄
MW: 243 g/mol
Density: 1.7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01631569
CAS Number: 261951-85-3