2-Fluoro-4-hydroxybenzonitrile 99%
Supplier: Apollo Scientific
Danger
PC0409-100G PC0409-25G PC0409-5G
APOSPC0409-100GEA 82.8 EUR
APOSPC0409-100G APOSPC0409-25G APOSPC0409-5G
2-Fluoro-4-hydroxybenzonitrile 99%
2-Fluoro-4-hydroxybenzonitrile
Formula: C₇H₄FNO
MW: 137.11 g/mol
Melting Pt: 123…126 °C
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00192177
CAS Number: 82380-18-5
EINECS: 422-810-7
UN: 3077
ADR: 9+EHS,III