1,2-Dibromo-1-chloro-2,2-difluoroethane 97%
Supplier: Apollo Scientific
Synonyms: 1-Chloro-1,2-dibromo-2,2-difluoroethane
PC0421-1G PC0421-5G
APOSPC0421-1GEA 107 EUR
APOSPC0421-1G APOSPC0421-5G
1,2-Dibromo-1-chloro-2,2-difluoroethane 97%
1,2-Dibromo-1-chloro-2,2-difluoroethane
Formula: C₂HBr₂ClF₂
MW: 258.3 g/mol
Boiling Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD01075262
CAS Number: 421-36-3