2-Methyl-5-nitrobenzotrifluoride
Supplier: Apollo Scientific
Danger
Synonyms: 1-Methyl-4-nitro-2-(trifluoromethyl)benzene
PC0422-25G PC0422-5G PC0422-100G
APOSPC0422-25GEA 58 EUR
APOSPC0422-25G APOSPC0422-5G APOSPC0422-100G
2-Methyl-5-nitrobenzotrifluoride
2-Methyl-5-nitrobenzotrifluoride
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631684
CAS Number: 89976-12-5
UN: 3439
ADR: 6.1 III