2-Methyl-5-(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
Warning
PC0447-5G PC0447-1G PC0447-10G
APOSPC0447-5GEA 40.6 EUR
APOSPC0447-5G APOSPC0447-1G APOSPC0447-10G
2-Methyl-5-(trifluoromethyl)benzoic acid 97%
2-Methyl-5-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 123…125 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631589
CAS Number: 13055-63-5