2-Methyl-3-(trifluoromethyl)benzyl alcohol 98%
Supplier: Apollo Scientific
PC0470-1G PC0470-5G PC0470-25G
APOSPC0470-1GEA 56.7 EUR
APOSPC0470-1G APOSPC0470-5G APOSPC0470-25G
2-Methyl-3-(trifluoromethyl)benzyl alcohol 98%
2-Methyl-3-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O
MW: 190.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631603
CAS Number: 261952-12-9