3-Nitrophenylsulphur pentafluoride 96+%
Supplier: Apollo Scientific
Warning
PC0472-1G PC0472-25G PC0472-5G
APOSPC0472-1GEA 335 EUR
APOSPC0472-1G APOSPC0472-25G APOSPC0472-5G
3-Nitrophenylsulphur pentafluoride 96+%
3-Nitrophenylsulphur pentafluoride
Formula: C₆H₄F₅NO₂S
MW: 249.16 g/mol
Boiling Pt: 106 °C/2mmHg
Density: 1.662 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00221621
CAS Number: 2613-26-5
EINECS: 447-600-2
UN: 2810
ADR: 6.1 III