6-(Trifluoromethyl)nicotinic acid 98%
Supplier: Apollo Scientific
Warning
PC0481-1G PC0481-25G PC0481-5G
APOSPC0481-1GEA 21.9 EUR
APOSPC0481-1G APOSPC0481-25G APOSPC0481-5G
6-(Trifluoromethyl)nicotinic acid 98%
6-(Trifluoromethyl)nicotinic acid
Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00792430
CAS Number: 231291-22-8