3,5-Difluoro-4-methoxybenzoic acid 99%
Supplier: Apollo Scientific
Warning
PC0494-1G PC0494-25G PC0494-5G
APOSPC0494-1GEA 21.9 EUR
APOSPC0494-1G APOSPC0494-25G APOSPC0494-5G
3,5-Difluoro-4-methoxybenzoic acid 99%
3,5-Difluoro-4-methoxybenzoic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 164…166 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01090994
CAS Number: 319-60-8