2,4,6-Trifluoroanisole 98%
Supplier: Apollo Scientific
Synonyms: 1,3,5-Trifluoro-2-methoxybenzene, 1-Methoxy-2,4,6-trifluorobenzene
PC0511-10G PC0511-5G PC0511-1G
APOSPC0511-10GEA 168 EUR
APOSPC0511-10G APOSPC0511-5G APOSPC0511-1G
2,4,6-Trifluoroanisole 98%
2,4,6-Trifluoroanisole
Formula: C₇H₅F₃O
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD01631502
CAS Number: 219998-30-8
UN: 3271
ADR: 3,III