2-(Trifluoromethoxy)anisole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Methoxy-2-(trifluoromethoxy)benzene, 2-Trifluoromethoxyanisole
PC0515-1G
PC0515-5G
PC0515-25G
APOSPC0515-1GEA
21.9
EUR
APOSPC0515-1G
APOSPC0515-5G
APOSPC0515-25G
2-(Trifluoromethoxy)anisole 97%
2-(Trifluoromethoxy)anisole
Formula:
C₈H₇F₃O₂ MW: 192.14 g/mol Boiling Pt: 69…70 °C/10mmHg Flash Pt: 47 °C Storage Temperature: Ambient |
CAS Number:
261952-22-1 UN: 1993 ADR: 3 III |