4-(Trifluoromethoxy)-DL-phenylglycine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-2-(4-(trifluoromethoxy)phenyl)acetic acid
PC0518-1G PC0518-250MG PC0518-5G PC0518-100MG
APOSPC0518-1GEA 174 EUR
APOSPC0518-1G APOSPC0518-250MG APOSPC0518-5G APOSPC0518-100MG
4-(Trifluoromethoxy)-DL-phenylglycine
4-(Trifluoromethoxy)-DL-phenylglycine
Formula: C₉H₈F₃NO₃
MW: 235.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631461
CAS Number: 261952-24-3